Gene Transcripts

Gene Symbol
WLS
Entrez Gene ID
79971
Gene Name
wntless Wnt ligand secretion mediator
Gene Aliases
C1orf139, EVI, GPR177, MRP, mig-14
Chromosome Location
Chr. 1: 68098459 - 68233000 on Build GRCh38
UniGene ID
Hs.647659
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002292.3 NP_001002292.3 5 945
NM_001193334.1 NP_001180263.1 4 678
NM_024911.6 NP_079187.3 5 1006
XM_011542191.2 XP_011540493.1 5 929
XM_011542192.2 XP_011540494.1 5 939
XM_017002390.1 XP_016857879.1 5 773
GenBank mRNA AK026744.1 - 2 87
AK074984.1 - 5 928
AK301613.1 - 4 538
AK309779.1 - 6 885
AY359035.1 - 5 988
BC007211.1 - 4 544
BC110826.1 - 5 907 6183768
BC137109.1 - 5 803 9021106
BC137113.1 - 5 803 9021110
BX538320.1 - 5 945
BX648748.1 - 4 676
DQ323735.1 - 5 975

View Full Product Details