Gene Transcripts

Gene Symbol
GREB1L
Entrez Gene ID
80000
Gene Name
growth regulation by estrogen in breast cancer 1 like
Gene Aliases
C18orf6, KIAA1772
Chromosome Location
Chr. 18: 21242225 - 21526273 on Build GRCh38
UniGene ID
Hs.149020
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142966.1 NP_001136438.1 25 4518
XM_006722547.2 XP_006722610.1 25 4481
XM_011526179.2 XP_011524481.1 25 4481
XM_017025988.1 XP_016881477.1 25 4481
XM_017025989.1 XP_016881478.1 25 4554
XM_017025990.1 XP_016881479.1 25 4481
XM_017025991.1 XP_016881480.1 25 4397
XM_017025992.1 XP_016881481.1 24 4352
XM_017025993.1 XP_016881482.1 24 4425
XM_017025994.1 XP_016881483.1 24 4154
XM_017025995.1 XP_016881484.1 24 4070
XM_017025996.1 XP_016881485.1 23 4025
GenBank mRNA AK023749.1 - 12 2271
AK293321.1 - 13 2465

View Full Product Details