Gene Transcripts
- Gene Symbol
- KLHL18
- Entrez Gene ID
- 23276
- Gene Name
- kelch like family member 18
- Gene Aliases
- Chromosome Location
- Chr. 3: 47282840 - 47346816 on Build GRCh38
- UniGene ID
- Hs.517946
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025010.4 |
NP_079286.2
|
4, 5 |
728 |
|
XM_005265001.3 |
XP_005265058.2
|
4, 5 |
628 |
|
XM_005265002.4 |
XP_005265059.2
|
4, 5 |
628 |
|
XM_005265003.3 |
XP_005265060.2
|
4, 5 |
628 |
|
XM_006713079.1 |
XP_006713142.1
|
4, 5 |
584 |
|
XM_017006033.1 |
XP_016861522.1
|
3, 4 |
491 |
|
XM_017006034.1 |
XP_016861523.1
|
3, 4 |
491 |
|
XM_017006035.1 |
XP_016861524.1
|
3, 4 |
491 |
|
GenBank mRNA |
AB018338.1 |
- |
2, 3 |
276 |
|
AB062478.1 |
- |
4, 5 |
900 |
|
AK023765.1 |
- |
4, 5 |
623 |
|
AK291477.1 |
- |
4, 5 |
622 |
|
AK295292.1 |
- |
3, 4 |
490 |
|
AY714879.1 |
- |
1, 2 |
155 |
|
BC032620.1 |
- |
4, 5 |
540 |
5578173 |
BX537953.1 |
- |
4, 5 |
619 |
|
View Full Product Details