Gene Transcripts
- Gene Symbol
- MAPK13
- Entrez Gene ID
- 5603
- Gene Name
- mitogen-activated protein kinase 13
- Gene Aliases
- MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta
- Chromosome Location
- Chr. 6: 36130484 - 36144524 on Build GRCh38
- UniGene ID
- Hs.178695
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002754.4 |
NP_002745.1
|
4, 5 |
516 |
|
NR_072996.1 |
|
4, 5 |
516 |
|
GenBank mRNA |
AB209586.1 |
- |
3, 4 |
1474 |
|
AB451221.1 |
- |
5, 6 |
417 |
|
AB451344.1 |
- |
5, 6 |
417 |
|
AF004709.1 |
- |
4, 5 |
465 |
|
AF015256.1 |
- |
4, 5 |
477 |
|
AF092535.1 |
- |
4, 5 |
438 |
|
AF100546.1 |
- |
4, 5 |
419 |
|
AK222922.1 |
- |
4, 5 |
408 |
|
AK308018.1 |
- |
4, 5 |
497 |
|
BC000433.2 |
- |
4, 5 |
458 |
2819932 |
BC001641.2 |
- |
4, 5 |
458 |
2819932 |
BC004428.1 |
- |
4, 5 |
458 |
2819932 |
BC085196.1 |
- |
4, 5 |
486 |
6107307 |
BT007221.1 |
- |
4, 5 |
417 |
|
CR536490.1 |
- |
4, 5 |
417 |
|
CR541687.1 |
- |
4, 5 |
417 |
|
DB473379.1 |
- |
|
480 |
|
DR006449.1 |
- |
4, 5 |
520 |
|
KU178281.1 |
- |
4, 5 |
417 |
|
KU178282.1 |
- |
4, 5 |
417 |
|
U93232.1 |
- |
4, 5 |
424 |
|
Y10488.1 |
- |
4, 5 |
439 |
|
View Full Product Details