Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190274.1 | NP_001177203.1 | 4 | 549 | |
NM_025133.4 | NP_079409.3 | 4 | 338 | ||
XM_005264572.4 | XP_005264629.1 | 4 | 543 | ||
XM_005264573.4 | XP_005264630.1 | 4 | 496 | ||
XM_017005015.1 | XP_016860504.1 | 4 | 496 | ||
XM_017005016.1 | XP_016860505.1 | 4 | 362 | ||
XM_017005017.1 | XP_016860506.1 | 4 | 362 | ||
GenBank mRNA | AF174599.1 | - | 4 | 431 | |
AF176706.1 | - | 4 | 419 | ||
AF351618.1 | - | 4 | 351 | ||
AK223592.1 | - | 4 | 433 | ||
AK292877.1 | - | 4 | 388 | ||
AK297444.1 | - | 4 | 523 | ||
AK304140.1 | - | 4 | 253 | ||
AK315108.1 | - | 4 | 338 | ||
AY827075.1 | - | 4 | 476 | ||
BC043258.1 | - | 4 | 440 | ||
BC130445.1 | - | 4 | 274 | 40146476 | |
BC136480.1 | - | 4 | 274 | 9020469 | |
DN994547.1 | - | 4 | 557 |