Gene Transcripts

Gene Symbol
NRBF2
Entrez Gene ID
29982
Gene Name
nuclear receptor binding factor 2
Gene Aliases
COPR, COPR1, COPR2, NRBF-2
Chromosome Location
Chr. 10: 63133247 - 63155031 on Build GRCh38
UniGene ID
Hs.449628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030759.4 NP_110386.2 2 305
XM_006717810.3 XP_006717873.1 2 267
GenBank mRNA AF267866.1 - 2 256
AK054957.1 - 2 263
AK225256.1 - 2 214
AK225257.1 - 2 216
AL136656.1 - 2 260
AU310836.1 - 2 305
AY267839.1 - 2 193
AY267840.1 - 2 193
BC011707.1 - 2 179 3687848

View Full Product Details