Gene Transcripts

Gene Symbol
MRPS26
Entrez Gene ID
64949
Gene Name
mitochondrial ribosomal protein S26
Gene Aliases
C20orf193, GI008, MRP-S13, MRP-S26, MRPS13, NY-BR-87, RPMS13, dJ534B8.3
Chromosome Location
Chr. 20: 3046029 - 3048250 on Build GRCh38
UniGene ID
Hs.18946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030811.3 NP_110438.1 4 537
GenBank mRNA AF308301.1 - 4 488
BC013018.2 - 4 504 3051126
BI115733.1 - 4 621

View Full Product Details