Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001306199.1 | NP_001293128.1 | 6 | 1325 | |
NM_030648.3 | NP_085151.1 | 6 | 1325 | ||
XM_017008661.1 | XP_016864150.1 | 4 | 418 | ||
GenBank mRNA | AB051504.1 | - | 6 | 960 | |
AF448510.1 | - | 6 | 960 | ||
AF462150.1 | - | 6 | 693 | ||
AK310856.1 | - | 6 | 794 | ||
BC121055.1 | - | 6 | 869 | 40118363 | |
BC121056.1 | - | 6 | 869 | 40118365 |