Gene Transcripts

Gene Symbol
KAZALD1
Entrez Gene ID
81621
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
BONO1, FKSG28, FKSG40, IGFBP-rP10
Chromosome Location
Chr. 10: 101061141 - 101068141 on Build GRCh38
UniGene ID
Hs.534859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319303.1 NP_001306232.1 3 1549
NM_030929.4 NP_112191.2 3 1494
NR_135067.1 2 932
XM_005270194.3 XP_005270251.1 3 1595
XM_017016714.1 XP_016872203.1 2 573
XM_017016715.1 XP_016872204.1 2 573
GenBank mRNA AK172862.1 - 3 745
AK172864.1 - 3 1548
AK225949.1 - 3 746
AY359087.1 - 3 750
BC007758.2 - 3 895 4122366
BI766345.1 - 2 205
CX871488.1 - 2 223

View Full Product Details