Gene Transcripts

Gene Symbol
CALN1
Entrez Gene ID
83698
Gene Name
calneuron 1
Gene Aliases
CABP8
Chromosome Location
Chr. 7: 71779491 - 72504271 on Build GRCh38
UniGene ID
Hs.592750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017440.2 NP_001017440.1 2, 3 643
NM_031468.3 NP_113656.2 3, 4 633
XM_011516594.2 XP_011514896.1 2, 3 485
XM_011516595.1 XP_011514897.1 2, 3 233
XM_011516596.2 XP_011514898.1 3, 4 1128
XM_011516597.2 XP_011514899.1 2, 3 1061
XM_011516599.1 XP_011514901.1 2, 3 432
XM_017012675.1 XP_016868164.1 4, 5 988
XM_017012676.1 XP_016868165.1 3, 4 770
XM_017012677.1 XP_016868166.1 2, 3 283
XM_017012678.1 XP_016868167.1 3, 4 745
XM_017012679.1 XP_016868168.1 2, 3 1098
XM_017012680.1 XP_016868169.1 2, 3 397
XM_017012681.1 XP_016868170.1 2, 3 260
XM_017012682.1 XP_016868171.1 2, 3 810
XM_017012683.1 XP_016868172.1 3, 4 594
GenBank mRNA AF282250.1 - 3, 4 364
AK123862.1 - 2, 3 178
AK313054.1 - 2, 3 233
AY007302.1 - 2, 3 291
AY726595.1 - 1, 2 119
BC020200.1 - 2, 3 415 3944302
BG718538.1 - 3, 4 638

View Full Product Details