Gene Transcripts

Gene Symbol
SLC25A18
Entrez Gene ID
83733
Gene Name
solute carrier family 25 member 18
Gene Aliases
GC2
Chromosome Location
Chr. 22: 17562470 - 17590890 on Build GRCh38
UniGene ID
Hs.282982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303484.1 NP_001290413.1 3, 4 622
NM_031481.2 NP_113669.1 4, 5 685
XM_011546149.2 XP_011544451.1 4, 5 1614
XM_011546150.2 XP_011544452.1 4, 5 1484
XM_011546151.2 XP_011544453.1 4, 5 873
XM_011546152.1 XP_011544454.1 4, 5 500
XM_011546153.2 XP_011544455.1 4, 5 1495
XM_011546154.2 XP_011544456.1 4, 5 742
XM_017028968.1 XP_016884457.1 4, 5 368
XM_017028969.1 XP_016884458.1 4, 5 2514
XM_017028970.1 XP_016884459.1 5, 6 2005
XM_017028971.1 XP_016884460.1 5, 6 1994
XM_017028972.1 XP_016884461.1 5, 6 1917
XM_017028973.1 XP_016884462.1 4, 5 1942
XM_017028974.1 XP_016884463.1 5, 6 1873
XM_017028975.1 XP_016884464.1 5, 6 1785
GenBank mRNA AJ428203.1 - 4, 5 641
AK095706.1 - 3, 4 1607
AK309110.1 - 3, 4 469
AY008285.1 - 4, 5 641
BC031644.1 - 3, 4 577 5179038
CR456578.1 - 2, 3 271

View Full Product Details