Gene Transcripts

Gene Symbol
INHBE
Entrez Gene ID
83729
Gene Name
inhibin beta E subunit
Gene Aliases
Chromosome Location
Chr. 12: 57455313 - 57458008 on Build GRCh38
UniGene ID
Hs.632713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031479.3 NP_113667.1 1 505
GenBank mRNA AF412024.1 - 1 489
AK075285.1 - 1 500
BC005161.2 - 1 497 3529092

View Full Product Details