Gene Transcripts

Gene Symbol
SLC25A18
Entrez Gene ID
83733
Gene Name
solute carrier family 25 member 18
Gene Aliases
GC2
Chromosome Location
Chr. 22: 17562470 - 17590890 on Build GRCh38
UniGene ID
Hs.282982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303484.1 NP_001290413.1 7 935
NM_031481.2 NP_113669.1 8 998
XM_011546149.2 XP_011544451.1 8 1927
XM_011546150.2 XP_011544452.1 8 1797
XM_011546151.2 XP_011544453.1 8 1186
XM_011546152.1 XP_011544454.1 8 813
XM_011546153.2 XP_011544455.1 8 1808
XM_011546154.2 XP_011544456.1 8 1055
XM_017028968.1 XP_016884457.1 8 681
XM_017028969.1 XP_016884458.1 8 2827
XM_017028970.1 XP_016884459.1 9 2318
XM_017028971.1 XP_016884460.1 9 2307
XM_017028972.1 XP_016884461.1 9 2230
XM_017028973.1 XP_016884462.1 8 2255
XM_017028974.1 XP_016884463.1 9 2186
XM_017028975.1 XP_016884464.1 9 2098
GenBank mRNA AJ428203.1 - 8 954
AK309110.1 - 7 782
AY008285.1 - 8 954
BC031644.1 - 7 890 5179038
CR456578.1 - 6 584

View Full Product Details