Gene Transcripts
- Gene Symbol
- ANO3
- Entrez Gene ID
- 63982
- Gene Name
- anoctamin 3
- Gene Aliases
- C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
- Chromosome Location
- Chr. 11: 26189123 - 26663289 on Build GRCh38
- UniGene ID
- Hs.577269
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001313726.1 |
NP_001300655.1
|
7, 8 |
937 |
|
NM_001313727.1 |
NP_001300656.1
|
3, 4 |
495 |
|
NM_031418.3 |
NP_113606.2
|
6, 7 |
845 |
|
XM_017018118.1 |
XP_016873607.1
|
4, 5 |
473 |
|
GenBank mRNA |
AJ300461.1 |
- |
6, 7 |
1552 |
|
AK295143.1 |
- |
3, 4 |
408 |
|
AK295816.1 |
- |
3, 4 |
407 |
|
AK304781.1 |
- |
7, 8 |
778 |
|
AK316018.1 |
- |
3, 4 |
407 |
|
BP219022.1 |
- |
3, 4 |
496 |
|
View Full Product Details