Gene Transcripts

Gene Symbol
C1QTNF7
Entrez Gene ID
114905
Gene Name
C1q and tumor necrosis factor related protein 7
Gene Aliases
CTRP7, ZACRP7
Chromosome Location
Chr. 4: 15339818 - 15446169 on Build GRCh38
UniGene ID
Hs.153714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135170.1 NP_001128642.1 2 472
NM_001135171.1 NP_001128643.1 2 419
NM_031911.4 NP_114117.1 2 302
XM_011513772.1 XP_011512074.1 3 342
XM_011513773.1 XP_011512075.1 3 391
GenBank mRNA AF329839.1 - 2 277
AK314802.1 - 2 420
BC022187.2 - 2 420 4825040
BC024015.2 - 2 420 4823626
BX647781.1 - 2 472
DB069980.1 - 2 420
DB290077.1 - 2 303

View Full Product Details