Gene Transcripts

Gene Symbol
HDAC10
Entrez Gene ID
83933
Gene Name
histone deacetylase 10
Gene Aliases
HD10
Chromosome Location
Chr. 22: 50245183 - 50251405 on Build GRCh38
UniGene ID
Hs.26593
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032019.5 NP_114408.3 9 1168
GenBank mRNA AF393962.1 - 9 795
AF407272.1 - 9 795
AF426160.1 - 9 1148
BC032049.1 - 9 981
BC094734.1 - 8 943
BC125083.1 - 9 842 40117772
CR456465.1 - 9 851

View Full Product Details