Gene Transcripts
- Gene Symbol
- MND1
- Entrez Gene ID
- 84057
- Gene Name
- meiotic nuclear divisions 1
- Gene Aliases
- GAJ
- Chromosome Location
- Chr. 4: 153344649 - 153415097 on Build GRCh38
- UniGene ID
- Hs.294088
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253861.1 |
NP_001240790.1
|
3, 4 |
218 |
|
NM_032117.3 |
NP_115493.1
|
3, 4 |
218 |
|
NR_045605.1 |
|
3, 4 |
218 |
|
XM_005263275.2 |
XP_005263332.1
|
3, 4 |
177 |
|
XM_017008688.1 |
XP_016864177.1
|
3, 4 |
575 |
|
GenBank mRNA |
AK312172.1 |
- |
3, 4 |
177 |
|
AY028916.1 |
- |
3, 4 |
157 |
|
BC032142.2 |
- |
3, 4 |
172 |
5095005 |
BF975093.1 |
- |
3, 4 |
187 |
|
BG613292.1 |
- |
3, 4 |
180 |
|
BG758047.1 |
- |
3, 4 |
187 |
|
BM820228.1 |
- |
3, 4 |
218 |
|
BM833450.1 |
- |
3, 4 |
218 |
|
View Full Product Details