Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146160.1 | NP_001139632.1 | 2 | 85 | |
NM_001317889.1 | NP_001304818.1 | 4 | 201 | ||
NM_001317891.1 | NP_001304820.1 | 3 | 135 | ||
NM_032026.3 | NP_114415.1 | 4 | 201 | ||
NR_027427.1 | 4 | 201 | |||
XM_006716666.3 | XP_006716729.1 | 4 | 221 | ||
XM_006716667.3 | XP_006716730.1 | 4 | 190 | ||
XM_006716668.3 | XP_006716731.1 | 4 | 203 | ||
XM_006716669.3 | XP_006716732.1 | 2 | 87 | ||
XM_006716670.3 | XP_006716733.1 | 3 | 122 | ||
XM_006716671.3 | XP_006716734.1 | 3 | 124 | ||
XM_011517331.2 | XP_011515633.1 | 3 | 117 | ||
XM_017013895.1 | XP_016869384.1 | 4 | 211 | ||
XM_017013896.1 | XP_016869385.1 | 4 | 190 | ||
XM_017013897.1 | XP_016869386.1 | 3 | 111 | ||
XM_017013898.1 | XP_016869387.1 | 3 | 124 | ||
XM_017013899.1 | XP_016869388.1 | 4 | 197 | ||
GenBank mRNA | AF212250.1 | - | 4 | 184 | |
AK312204.1 | - | 4 | 197 | ||
AY071865.1 | - | 2 | 74 | ||
BC064964.1 | - | 4 | 185 | 6141834 | |
BG529611.1 | - | 3 | 126 | ||
BX332564.2 | - | 4 | 183 |