Gene Transcripts

Gene Symbol
IFI27L2
Entrez Gene ID
83982
Gene Name
interferon alpha inducible protein 27 like 2
Gene Aliases
FAM14A, ISG12B, TLH29
Chromosome Location
Chr. 14: 94127781 - 94129620 on Build GRCh38
UniGene ID
Hs.94695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032036.2 NP_114425.1 4 388
GenBank mRNA AF208232.1 - 4 354
AF238978.1 - 2 221
BC022800.2 - 2 296
BC032626.1 - 4 347 5229498

View Full Product Details