Gene Transcripts

Gene Symbol
MRO
Entrez Gene ID
83876
Gene Name
maestro
Gene Aliases
B29, C18orf3
Chromosome Location
Chr. 18: 50795120 - 50825413 on Build GRCh38
UniGene ID
Hs.30495
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB042647.1 - 5, 6 579
AK054702.1 - 5, 6 649
BC029860.1 - 5, 6 602 5171860

View Full Product Details