Gene Transcripts
- Gene Symbol
- NSRP1
- Entrez Gene ID
- 84081
- Gene Name
- nuclear speckle splicing regulatory protein 1
- Gene Aliases
- CCDC55, HSPC095, NSrp70
- Chromosome Location
- Chr. 17: 30115526 - 30186475 on Build GRCh38
- UniGene ID
- Hs.462663
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001261467.1 |
NP_001248396.1
|
2, 3 |
110 |
|
NM_032141.3 |
NP_115517.1
|
3, 4 |
204 |
|
XM_011525345.2 |
XP_011523647.1
|
3, 4 |
119 |
|
XM_017025209.1 |
XP_016880698.1
|
4, 5 |
323 |
|
XM_017025210.1 |
XP_016880699.1
|
3, 4 |
1034 |
|
XM_017025211.1 |
XP_016880700.1
|
4, 5 |
232 |
|
XM_017025212.1 |
XP_016880701.1
|
3, 4 |
369 |
|
XM_017025213.1 |
XP_016880702.1
|
2, 3 |
149 |
|
XM_017025214.1 |
XP_016880703.1
|
2, 3 |
178 |
|
XM_017025215.1 |
XP_016880704.1
|
2, 3 |
182 |
|
GenBank mRNA |
AF161358.1 |
- |
3, 4 |
183 |
|
AK094552.1 |
- |
|
1548 |
|
AK297102.1 |
- |
2, 3 |
104 |
|
AK313963.1 |
- |
3, 4 |
191 |
|
AL136806.1 |
- |
3, 4 |
195 |
|
BC016313.1 |
- |
4, 5 |
218 |
|
BC040118.1 |
- |
3, 4 |
184 |
5176197 |
BC046116.2 |
- |
3, 4 |
206 |
|
BC062425.1 |
- |
3, 4 |
196 |
|
BC105044.1 |
- |
3, 4 |
191 |
8144047 |
BC105046.1 |
- |
3, 4 |
191 |
8144049 |
BC143539.1 |
- |
2, 3 |
97 |
|
CB957325.1 |
- |
4, 5 |
294 |
|
CR533555.1 |
- |
3, 4 |
167 |
|
DC327004.1 |
- |
2, 3 |
110 |
|
View Full Product Details