Gene Transcripts
- Gene Symbol
- ANKRD17
- Entrez Gene ID
- 26057
- Gene Name
- ankyrin repeat domain 17
- Gene Aliases
- GTAR, MASK2, NY-BR-16
- Chromosome Location
- Chr. 4: 73073966 - 73259142 on Build GRCh38
- UniGene ID
- Hs.601206
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015574.1 |
NP_056389.1
|
1, 2 |
522 |
|
NM_032217.4 |
NP_115593.3
|
1, 2 |
522 |
|
NM_198889.2 |
NP_942592.1
|
1, 2 |
522 |
|
XM_005265671.3 |
XP_005265728.1
|
1, 2 |
866 |
|
XM_005265672.3 |
XP_005265729.1
|
1, 2 |
866 |
|
XM_005265673.3 |
XP_005265730.1
|
1, 2 |
866 |
|
XM_017008014.1 |
XP_016863503.1
|
1, 2 |
866 |
|
XM_017008016.1 |
XP_016863505.1
|
1, 2 |
866 |
|
GenBank mRNA |
AB014597.2 |
- |
1, 2 |
46 |
|
AF308285.1 |
- |
1, 2 |
509 |
|
AK307581.1 |
- |
1, 2 |
522 |
|
BC004173.2 |
- |
1, 2 |
503 |
|
BC043394.1 |
- |
1, 2 |
491 |
|
View Full Product Details