Gene Transcripts
- Gene Symbol
- BTBD10
- Entrez Gene ID
- 84280
- Gene Name
- BTB domain containing 10
- Gene Aliases
- GMRP-1, GMRP1
- Chromosome Location
- Chr. 11: 13387996 - 13463297 on Build GRCh38
- UniGene ID
- Hs.332382
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001297741.1 |
NP_001284670.1
|
7, 8 |
1230 |
|
NM_001297742.1 |
NP_001284671.1
|
7, 8 |
1420 |
|
NM_032320.6 |
NP_115696.2
|
8, 9 |
1388 |
|
XM_017018405.1 |
XP_016873894.1
|
8, 9 |
2504 |
|
XM_017018406.1 |
XP_016873895.1
|
9, 10 |
1483 |
|
XM_017018407.1 |
XP_016873896.1
|
9, 10 |
1463 |
|
XM_017018408.1 |
XP_016873897.1
|
8, 9 |
1358 |
|
XM_017018409.1 |
XP_016873898.1
|
8, 9 |
1330 |
|
XM_017018410.1 |
XP_016873899.1
|
8, 9 |
1322 |
|
XM_017018411.1 |
XP_016873900.1
|
7, 8 |
1217 |
|
GenBank mRNA |
AF329102.2 |
- |
8, 9 |
1388 |
|
AK095946.1 |
- |
7, 8 |
1228 |
|
AK292392.1 |
- |
8, 9 |
1313 |
|
AK294259.1 |
- |
7, 8 |
1209 |
|
AK298226.1 |
- |
7, 8 |
1105 |
|
AK316163.1 |
- |
7, 8 |
1420 |
|
AL832981.1 |
- |
8, 9 |
1369 |
|
AY221959.2 |
- |
8, 9 |
1431 |
|
BC005071.1 |
- |
8, 9 |
1350 |
3531347 |
BC017072.1 |
- |
3, 4 |
365 |
|
BC062633.1 |
- |
4, 5 |
498 |
|
View Full Product Details