Gene Transcripts

Gene Symbol
GON7
Entrez Gene ID
84520
Gene Name
GON7, KEOPS complex subunit homolog
Gene Aliases
C14orf142, PNAS-127
Chromosome Location
Chr. 14: 93202892 - 93207114 on Build GRCh38
UniGene ID
Hs.20142
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032490.4 NP_115879.2 2 879
GenBank mRNA BC021733.2 - 2 859
BC065216.1 - 2 781
BC107429.1 - 2 847 4809642
BC109390.1 - 2 790

View Full Product Details