Gene Transcripts

Gene Symbol
KBTBD8
Entrez Gene ID
84541
Gene Name
kelch repeat and BTB domain containing 8
Gene Aliases
TA-KRP, TAKRP
Chromosome Location
Chr. 3: 66998303 - 67011210 on Build GRCh38
UniGene ID
Hs.116665
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032505.2 NP_115894.2 3 556
XM_005264771.3 XP_005264828.1 2 341
GenBank mRNA AB058745.1 - 1 279
AF385438.1 - 2 489
AK096640.1 - 3 552
BC117487.1 - 2 453 40126038
DA246470.1 - 3 552

View Full Product Details