Gene Transcripts

Gene Symbol
PLEKHA8
Entrez Gene ID
84725
Gene Name
pleckstrin homology domain containing A8
Gene Aliases
FAPP2
Chromosome Location
Chr. 7: 30028361 - 30130483 on Build GRCh38
UniGene ID
Hs.233495
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032639.3 NP_116028.1 13 1799
XM_011515592.2 XP_011513894.1 14 1809
GenBank mRNA BC002838.2 - 13 1488

View Full Product Details