Gene Transcripts

Gene Symbol
RELT
Entrez Gene ID
84957
Gene Name
RELT tumor necrosis factor receptor
Gene Aliases
TNFRSF19L, TRLT
Chromosome Location
Chr. 11: 73376360 - 73397474 on Build GRCh38
UniGene ID
Hs.533720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032871.3 NP_116260.2 8 961
NM_152222.1 NP_689408.1 8 899
XM_005274356.2 XP_005274413.1 3 1062
XM_006718714.2 XP_006718777.1 6 526
XM_011545305.1 XP_011543607.1 7 784
GenBank mRNA AF319553.1 - 8 961
AK027899.1 - 8 899
AK057036.1 - 2 703
AK074128.1 - 1 2282
AK310103.1 - 5 767
BC017279.2 - 8 824 4873951
BC051810.1 - 8 851 5216988

View Full Product Details