Gene Transcripts

Gene Symbol
TMEM87B
Entrez Gene ID
84910
Gene Name
transmembrane protein 87B
Gene Aliases
Chromosome Location
Chr. 2: 112055223 - 112119318 on Build GRCh38
UniGene ID
Hs.656298
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032824.2 NP_116213.1 4 809
XM_005263827.2 XP_005263884.1 4 766
XM_006712807.2 XP_006712870.1 4 766
XM_017005121.1 XP_016860610.1 4 312
GenBank mRNA AK027587.1 - 4 794
AK074243.1 - 1 70
AK290294.1 - 4 455
AY429534.1 - 4 312
AY429535.1 - 4 312
BC115373.1 - 4 613 40026807

View Full Product Details