Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042616.2 | NP_001036081.1 | 2 | 607 | |
GenBank mRNA | AB206972.1 | - | 2 | 579 | |
BC007876.2 | - | 2 | 579 | 3947944 | |
BI548853.1 | - | 2 | 613 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032906.4 | NP_116295.1 | 2 | 607 | |
GenBank mRNA | BG706896.1 | - | 2 | 612 | |
CV812938.1 | - | 2 | 543 |