Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033123.3 | NP_149114.2 | 3, 4 | 409 | |
XM_005253519.2 | XP_005253576.1 | 3, 4 | 402 | ||
XM_017020176.1 | XP_016875665.1 | 3, 4 | 402 | ||
XM_017020177.1 | XP_016875666.1 | 3, 4 | 402 | ||
XM_017020178.1 | XP_016875667.1 | 3, 4 | 402 | ||
XM_017020181.1 | XP_016875670.1 | 3, 4 | 356 | ||
GenBank mRNA | AF532185.1 | - | 3, 4 | 334 | |
AK292279.1 | - | 3, 4 | 407 |