Gene Transcripts

Gene Symbol
GAL3ST3
Entrez Gene ID
89792
Gene Name
galactose-3-O-sulfotransferase 3
Gene Aliases
GAL3ST-3, GAL3ST2
Chromosome Location
Chr. 11: 66040765 - 66049180 on Build GRCh38
UniGene ID
Hs.208343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033036.2 NP_149025.1 3 1373
XM_017018519.1 XP_016874008.1 2 1287
GenBank mRNA AB053232.1 - 3 1373
AY026481.1 - 2 1251
BC113552.1 - 2 1159 8322604
BC113554.1 - 2 1159 8322606

View Full Product Details