Gene Transcripts

Gene Symbol
CPXCR1
Entrez Gene ID
53336
Gene Name
CPX chromosome region, candidate 1
Gene Aliases
CT77
Chromosome Location
Chr. X: 88747225 - 88754785 on Build GRCh38
UniGene ID
Hs.458292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184771.1 NP_001171700.1 3 406
NM_033048.5 NP_149037.4 3 444
GenBank mRNA AF284767.1 - 1 221
AK098646.1 - 3 381
AK313768.1 - 3 419
BC027614.2 - 3 414 4838752
BU570042.1 - 3 447

View Full Product Details