Gene Transcripts

Gene Symbol
RFX2
Entrez Gene ID
5990
Gene Name
regulatory factor X2
Gene Aliases
Chromosome Location
Chr. 19: 5993164 - 6110653 on Build GRCh38
UniGene ID
Hs.465709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000635.3 NP_000626.2 18 2603
NM_134433.2 NP_602309.1 17 2528
XM_011528171.2 XP_011526473.1 19 2569
XM_011528172.1 XP_011526474.1 18 2430
XM_017027107.1 XP_016882596.1 17 2453
XM_017027108.1 XP_016882597.1 16 2378
GenBank mRNA AK024288.1 - 15 2142
AK093977.1 - 10 3158
AK291196.1 - 17 2407
AK302661.1 - 12 1588
AK303321.1 - 15 1912
BC028579.1 - 18 2515 4829582
BC071571.1 - 18 2484 5276369
BX648130.1 - 18 2438
X76091.1 - 18 2493

View Full Product Details