Gene Transcripts

Gene Symbol
NKX3-2
Entrez Gene ID
579
Gene Name
NK3 homeobox 2
Gene Aliases
BAPX1, NKX3.2, NKX3B, SMMD
Chromosome Location
Chr. 4: 13540830 - 13544490 on Build GRCh38
UniGene ID
Hs.590927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001189.3 NP_001180.1 2 1894
GenBank mRNA AF009801.1 - 2 1818

View Full Product Details