Gene Transcripts
- Gene Symbol
- ARHGAP18
- Entrez Gene ID
- 93663
- Gene Name
- Rho GTPase activating protein 18
- Gene Aliases
- MacGAP, SENEX, bA307O14.2
- Chromosome Location
- Chr. 6: 129576145 - 129710225 on Build GRCh38
- UniGene ID
- Hs.486458
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033515.2 |
NP_277050.2
|
2, 3 |
415 |
|
XM_005267212.2 |
XP_005267269.2
|
2, 3 |
491 |
|
XM_005267213.1 |
XP_005267270.1
|
2, 3 |
415 |
|
XM_017011494.1 |
XP_016866983.1
|
2, 3 |
491 |
|
GenBank mRNA |
AB053293.1 |
- |
1, 2 |
213 |
|
AK098594.1 |
- |
2, 3 |
415 |
|
AK307796.1 |
- |
4, 5 |
490 |
|
AL834511.1 |
- |
2, 3 |
353 |
|
AY643712.1 |
- |
2, 3 |
416 |
|
BC017223.2 |
- |
2, 3 |
337 |
|
BC039611.1 |
- |
2, 3 |
344 |
|
BC062417.1 |
- |
2, 3 |
324 |
|
BC101708.1 |
- |
2, 3 |
409 |
8069214 |
BC107416.1 |
- |
2, 3 |
371 |
4040931 |
BC111940.1 |
- |
2, 3 |
409 |
8327408 |
BC143531.1 |
- |
2, 3 |
409 |
|
BM557223.1 |
- |
2, 3 |
352 |
|
CB149638.1 |
- |
2, 3 |
415 |
|
View Full Product Details