Gene Transcripts

Gene Symbol
MED8
Entrez Gene ID
112950
Gene Name
mediator complex subunit 8
Gene Aliases
ARC32
Chromosome Location
Chr. 1: 43383908 - 43389812 on Build GRCh38
UniGene ID
Hs.301756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001653.2 NP_001001653.1 5 570
NM_052877.4 NP_443109.2 5 527
NM_201542.4 NP_963836.2 5 527
GenBank mRNA AF521562.1 - 4 479
BC010019.2 - 5 542 2905623
BC010543.1 - 4 468
BG620658.1 - 5 528
BI488866.1 - 4 483

View Full Product Details