Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033467.3 | NP_258428.2 | 3 | 428 | |
XM_017002310.1 | XP_016857799.1 | 4 | 694 | ||
XM_017002311.1 | XP_016857800.1 | 3 | 428 | ||
XM_017002313.1 | XP_016857802.1 | 3 | 428 | ||
XM_017002314.1 | XP_016857803.1 | 3 | 428 | ||
XM_017002315.1 | XP_016857804.1 | 3 | 304 | ||
GenBank mRNA | AF336981.1 | - | 2 | 251 | |
AK093058.1 | - | 3 | 376 | ||
BC101027.2 | - | 2 | 224 | 40007698 | |
BC101028.1 | - | 2 | 224 | 40007699 | |
BC101029.2 | - | 2 | 224 | 40007702 | |
BC101030.1 | - | 2 | 224 | 40007706 | |
DB089501.1 | - | 3 | 428 |