Gene Transcripts

Gene Symbol
SEPT1
Entrez Gene ID
1731
Gene Name
septin 1
Gene Aliases
DIFF6, LARP, PNUTL3, SEP1
Chromosome Location
Chr. 16: 30378133 - 30382850 on Build GRCh38
UniGene ID
Hs.632176
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052838.4 NP_443070.5 8, 9 840
XM_017022991.1 XP_016878480.1 8, 9 820
GenBank mRNA AF085235.1 - 7, 8 671
AL833004.1 - 2, 3 1606
AY034176.1 - 7, 8 674
BC012161.1 - 6, 7 662 4576220

View Full Product Details