Gene Transcripts
- Gene Symbol
- CDCA5
- Entrez Gene ID
- 113130
- Gene Name
- cell division cycle associated 5
- Gene Aliases
- SORORIN
- Chromosome Location
- Chr. 11: 65061009 - 65084143 on Build GRCh38
- UniGene ID
- Hs.434886
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080668.3 |
NP_542399.1
|
4, 5 |
406 |
|
XM_005273733.3 |
XP_005273790.1
|
4, 5 |
401 |
|
XM_005273734.3 |
XP_005273791.1
|
4, 5 |
401 |
|
XM_011544743.2 |
XP_011543045.1
|
4, 5 |
401 |
|
XM_011544748.2 |
XP_011543050.1
|
4, 5 |
401 |
|
XM_011544749.2 |
XP_011543051.1
|
4, 5 |
401 |
|
XM_017017158.1 |
XP_016872647.1
|
4, 5 |
401 |
|
GenBank mRNA |
AK291490.1 |
- |
4, 5 |
330 |
|
AL832526.1 |
- |
3, 4 |
123 |
|
BC011000.2 |
- |
4, 5 |
314 |
3938081 |
CN332525.1 |
- |
4, 5 |
405 |
|
View Full Product Details