Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080752.3 | NP_542790.2 | 1 | 327 | |
NR_037628.1 | 1 | 327 | |||
GenBank mRNA | AK056641.1 | - | 1 | 292 | |
AK302767.1 | - | 1 | 291 | ||
AK316214.1 | - | 1 | 290 | ||
BC027611.2 | - | 1 | 284 | 4829980 | |
BX385024.2 | - | 1 | 285 | ||
DA520427.1 | - | 1 | 327 | ||
DC363945.1 | - | 1 | 290 |