Gene Transcripts

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
Chromosome Location
Chr. 7: 117363222 - 117427523 on Build GRCh38
UniGene ID
Hs.352412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301821.1 NP_001288750.1 3 375
NM_001301822.1 NP_001288751.1 3 375
NM_130768.2 NP_570124.1 3 375
GenBank mRNA AF461259.1 - 3 341
AK093445.1 - 3 328
BC028695.1 - 3 345
BC034963.1 - 3 350 4824818
BC126186.1 - 3 334 8991902
BC126188.1 - 3 334 8991904
BC144212.1 - 3 334 9052732
DB457846.1 - 3 375

View Full Product Details