Gene Transcripts

Gene Symbol
ZSWIM3
Entrez Gene ID
140831
Gene Name
zinc finger SWIM-type containing 3
Gene Aliases
C20orf164, PPP1R174
Chromosome Location
Chr. 20: 45857581 - 45879131 on Build GRCh38
UniGene ID
Hs.292135
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080752.3 NP_542790.2 2 418
NR_037628.1 3 573
GenBank mRNA AK056641.1 - 2 383
AK302767.1 - 3 537
AK316214.1 - 3 536
BC027611.2 - 2 375 4829980
BX385024.2 - 2 376
DA520427.1 - 2 418
DC363945.1 - 3 536

View Full Product Details