Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133367.4 | NP_588608.1 | 2 | 1425 | |
GenBank mRNA | AF313619.1 | - | 1 | 857 | |
AF347029.1 | - | 2 | 1415 | ||
AK093603.1 | - | 1 | 1041 | ||
AK128085.1 | - | 1 | 1645 | ||
AY424286.1 | - | 3 | 1428 | ||
BC030664.2 | - | 2 | 1400 | 5260230 |