Gene Transcripts

Gene Symbol
PHF21B
Entrez Gene ID
112885
Gene Name
PHD finger protein 21B
Gene Aliases
BHC80L, PHF4
Chromosome Location
Chr. 22: 44881162 - 45009928 on Build GRCh38
UniGene ID
Hs.254097
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138415.4 NP_612424.1 5 849
GenBank mRNA BC012187.1 - 5 849
KU178842.1 - 5 698

View Full Product Details