Gene Transcripts
- Gene Symbol
- SYTL5
- Entrez Gene ID
- 94122
- Gene Name
- synaptotagmin like 5
- Gene Aliases
- slp5
- Chromosome Location
- Chr. X: 37888915 - 38128820 on Build GRCh38
- UniGene ID
- Hs.662334
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163334.1 |
NP_001156806.1
|
1, 2 |
469 |
|
NM_001163335.1 |
NP_001156807.1
|
3, 4 |
647 |
|
NM_138780.2 |
NP_620135.1
|
2, 3 |
556 |
|
XM_011544001.2 |
XP_011542303.1
|
4, 5 |
908 |
|
XM_011544002.2 |
XP_011542304.1
|
2, 3 |
542 |
|
XM_017029972.1 |
XP_016885461.1
|
5, 6 |
1074 |
|
XM_017029973.1 |
XP_016885462.1
|
4, 5 |
1141 |
|
XM_017029974.1 |
XP_016885463.1
|
3, 4 |
769 |
|
GenBank mRNA |
AB080222.1 |
- |
1, 2 |
113 |
|
AK125106.1 |
- |
2, 3 |
525 |
|
BC131585.1 |
- |
1, 2 |
113 |
40113205 |
BX647688.1 |
- |
3, 4 |
649 |
|
View Full Product Details