Gene Transcripts

Gene Symbol
TM4SF18
Entrez Gene ID
116441
Gene Name
transmembrane 4 L six family member 18
Gene Aliases
L6D
Chromosome Location
Chr. 3: 149318498 - 149333761 on Build GRCh38
UniGene ID
Hs.22026
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184723.1 NP_001171652.1 5 1072
NM_138786.3 NP_620141.1 6 1248
GenBank mRNA AL833134.1 - 6 1170
BC014339.1 - 6 1124 3828466

View Full Product Details