Gene Transcripts
- Gene Symbol
- DPP9
- Entrez Gene ID
- 91039
- Gene Name
- dipeptidyl peptidase 9
- Gene Aliases
- DP9, DPLP9, DPRP-2, DPRP2
- Chromosome Location
- Chr. 19: 4675226 - 4723843 on Build GRCh38
- UniGene ID
- Hs.515081
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_139159.4 |
NP_631898.3
|
7, 8 |
1035 |
|
XM_005259673.2 |
XP_005259730.1
|
8, 9 |
1144 |
|
XM_011528405.1 |
XP_011526707.1
|
9, 10 |
1196 |
|
XM_011528406.1 |
XP_011526708.1
|
10, 11 |
1306 |
|
XM_011528408.1 |
XP_011526710.1
|
7, 8 |
981 |
|
XM_011528410.1 |
XP_011526712.1
|
8, 9 |
1196 |
|
XM_017027453.1 |
XP_016882942.1
|
9, 10 |
1786 |
|
XM_017027454.1 |
XP_016882943.1
|
10, 11 |
1896 |
|
GenBank mRNA |
AF452102.2 |
- |
7, 8 |
1055 |
|
AF542510.2 |
- |
7, 8 |
1014 |
|
AK122645.1 |
- |
1, 2 |
85 |
|
AK122654.1 |
- |
7, 8 |
1038 |
|
AK131100.1 |
- |
4, 5 |
604 |
|
AK131499.1 |
- |
6, 7 |
548 |
|
AL834376.1 |
- |
1, 2 |
66 |
|
AY172660.1 |
- |
4, 5 |
700 |
|
AY374518.1 |
- |
4, 5 |
690 |
|
BC037948.2 |
- |
7, 8 |
972 |
4577856 |
DQ417928.1 |
- |
5, 6 |
777 |
|
View Full Product Details