Gene Transcripts

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
Chromosome Location
Chr. 10: 70672518 - 70762441 on Build GRCh38
UniGene ID
Hs.352156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080722.4 NP_542453.2 2 755
NM_139155.3 NP_631894.2 2 755
XM_011539300.2 XP_011537602.1 2 739
XM_011539303.2 XP_011537605.1 2 743
XM_011539305.2 XP_011537607.1 2 743
XM_011539306.2 XP_011537608.1 2 743
XM_011539307.2 XP_011537609.1 2 743
XM_011539308.2 XP_011537610.1 2 743
GenBank mRNA AF358666.1 - 2 458
AF366351.1 - 1 376
AJ345098.1 - 2 458

View Full Product Details