Gene Transcripts
- Gene Symbol
- DPP9
- Entrez Gene ID
- 91039
- Gene Name
- dipeptidyl peptidase 9
- Gene Aliases
- DP9, DPLP9, DPRP-2, DPRP2
- Chromosome Location
- Chr. 19: 4675226 - 4723843 on Build GRCh38
- UniGene ID
- Hs.515081
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_139159.4 |
NP_631898.3
|
8, 9 |
1149 |
|
XM_005259673.2 |
XP_005259730.1
|
9, 10 |
1258 |
|
XM_011528405.1 |
XP_011526707.1
|
10, 11 |
1310 |
|
XM_011528406.1 |
XP_011526708.1
|
11, 12 |
1420 |
|
XM_011528408.1 |
XP_011526710.1
|
8, 9 |
1095 |
|
XM_011528410.1 |
XP_011526712.1
|
9, 10 |
1310 |
|
XM_017027453.1 |
XP_016882942.1
|
10, 11 |
1900 |
|
XM_017027454.1 |
XP_016882943.1
|
11, 12 |
2010 |
|
GenBank mRNA |
AF452102.2 |
- |
8, 9 |
1169 |
|
AF542510.2 |
- |
8, 9 |
1128 |
|
AK122645.1 |
- |
2, 3 |
199 |
|
AK122654.1 |
- |
8, 9 |
1152 |
|
AK131100.1 |
- |
5, 6 |
718 |
|
AK131499.1 |
- |
7, 8 |
662 |
|
AL834376.1 |
- |
2, 3 |
180 |
|
AY172660.1 |
- |
5, 6 |
814 |
|
AY374518.1 |
- |
5, 6 |
804 |
|
BC037948.2 |
- |
8, 9 |
1086 |
4577856 |
DQ417928.1 |
- |
6, 7 |
891 |
|
View Full Product Details