Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127715.2 | NP_001121187.1 | 24 | 2749 | |
NM_139244.4 | NP_640337.3 | 22 | 2641 | ||
XM_006715339.2 | XP_006715402.1 | 23 | 2875 | ||
XM_011535453.1 | XP_011533755.1 | 23 | 2473 | ||
XM_017010267.1 | XP_016865756.1 | 19 | 2820 | ||
GenBank mRNA | AB073398.1 | - | 13 | 1480 | |
AK055484.1 | - | 5 | 459 | ||
AK090549.1 | - | 14 | 1459 | ||
AK128121.1 | - | 23 | 2468 | ||
AL834152.1 | - | 25 | 2798 | ||
BC113382.1 | - | 22 | 2514 | 8322434 | |
BC113408.1 | - | 22 | 2514 | 8322460 |